missing translation for 'onlineSavingsMsg'
Learn More

Oclacitinib, TRC

Product Code. 30138678
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138678 5 mg 5mg
30135991 1 mg 1mg
30005882 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138678 Supplier Toronto Research Chemicals Supplier No. TRCO1700005MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1208319-26-9
Molecular Formula C15H23N5O2S
Molecular Weight (g/mol) 337.44
Synonym trans-N-Methyl-4-(methyl-7H-pyrrolo[2,3-d]pyrimidin-4-ylamino)cyclohexanemethanesulfonamide, JAKi, PF 03394197
IUPAC Name N-methyl-1-[4-[methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino]cyclohexyl]methanesulfonamide
SMILES CNS(=O)(=O)C[C@H]1CC[C@@H](CC1)N(C)C2=NC=NC3=C2C=CN3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H23N5O2S/c1-16-23(21,22)9-11-3-5-12(6-4-11)20(2)15-13-7-8-17-14(13)18-10-19-15/h7-8,10-12,16H,3-6,9H2,1-2H3,(H,17,18,19)/t11-,12-
Formula Weight 337.1572
Chemical Name or Material Oclacitinib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.