Learn More
Oct-3/4 Goat anti-Human, Polyclonal, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 29 publications
Brand:  R&D Systems AF1759-SP
Encompass
Product Code. 15761074
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Oct-3/4 Polyclonal specifically detects Oct-3/4 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.