missing translation for 'onlineSavingsMsg'
Learn More

Octacarbonyldicobalt, stab. with 1-5% hexane

Product Code. 11342148
Change view
Click to view available options
Quantity:
2.5 g
25 g
100 g
Unit Size:
100g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342148 25 g 25g
15431037 2.5 g 2.5g
11352148 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342148 Supplier Thermo Scientific Alfa Aesar Supplier No. 013060.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

As a catalyst in oxo process, and in high-purity cobalt salts | CAS: 10210-68-1

As a catalyst in oxo process, and in high-purity cobalt saltsOctacarbonyldicobalt is used as a catalyst for hydroformylation. It plays an important role in promoting both the Nicholas reaction and the Pauson-Khand reaction. It acts as a reagent and catalyst in organometallic chemistry and organic synthesis. It is involved as an alkyne protecting group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10210-68-1
MDL Number MFCD00016024
Synonym carbon monoxide; cobalt 2+, dicobalt octacarbonyl stabilised with hexane, cobalt carbonyl, moistened with hexane hexane co

Specifications

Melting Point ∼51°C (decomposition)
Color Red to Orange
Density 1.73 g/mL
Flash Point -13°C (-9°F)
Linear Formula Co2(CO)8
Quantity 25 g
UN Number UN3124
Sensitivity Air sensitive
Solubility Information Insoluble in water; Soluble in alcohol,ether and carbon disulfideSoluble in ether,naphtha and carbon disulfide; Slightly soluble in alcohol; Insoluble in water.
Formula Weight 341.95
Physical Form Crystalline
Chemical Name or Material Octacarbonyldicobalt, Stabilized with 1 to 5% hexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.