missing translation for 'onlineSavingsMsg'
Learn More

Octadecane, 99+%

Product Code. 11984391 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11984391 1 L Glass bottle 1 L
10625372 5 mL Glass bottle 5mL
10205420 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11984391 Supplier Thermo Scientific Acros Supplier No. 129290010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 593-45-3 | C18H38 | 254.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-45-3
Molecular Formula C18H38
Molecular Weight (g/mol) 254.5
InChI Key RZJRJXONCZWCBN-UHFFFAOYSA-N
Synonym n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group
PubChem CID 11635
ChEBI CHEBI:32926
IUPAC Name octadecane
SMILES CCCCCCCCCCCCCCCCCC

Specifications

Melting Point 28.0°C to 31.0°C
Color White
Density 0.7770g/mL
Boiling Point 317.0°C
Flash Point 166°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)16CH3
Quantity 1 L
Beilstein 01,173
Specific Gravity 0.777
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol, acetone and diethyl ether
Formula Weight 254.5
Percent Purity 99+%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Octadecane
Show More Show Less
compliance-icons
Product Identifier
  • Octadecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.