missing translation for 'onlineSavingsMsg'
Learn More

Octadecanedioic Acid, TRC

Product Code. 30414889
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30414889 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414889 Supplier Toronto Research Chemicals Supplier No. TRCO2353035G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octadecanedioic acid
CAS 871-70-5
Molecular Formula C18 H34 O4
Quantity 5 g
Synonym Octadecanedioic acid (6CI','7CI','8CI','9CI','ACI), 1,16-Dicarboxyhexadecane, 1,16-Hexadecanedicarboxylic acid, 1,18-Octadecanedioic acid, Emerox 118, Inherent C 18
InChI Formula InChI=1S/C18H34O4/c19-17(20)15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-16-18(21)22/h1-16H2,(H,19,20)(H,21,22)
SMILES OC(=O)CCCCCCCCCCCCCCCCC(=O)O
IUPAC Name octadecanedioic acid
Molecular Weight (g/mol) 314.46
Formula Weight 314.2457
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.