missing translation for 'onlineSavingsMsg'
Learn More

Octadecylbenzene, 98%

Product Code. 10319033 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10319033 50 g Glass bottle 50g
10091981 1 g Glass bottle 1g
10525291 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10319033 Supplier Thermo Scientific Acros Supplier No. 124700500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4445-07-2 | C24H42 | 330.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4445-07-2
Molecular Formula C24H42
Molecular Weight (g/mol) 330.59
MDL Number MFCD00048500
InChI Key WSVDSBZMYJJMSB-UHFFFAOYSA-N
IUPAC Name octadecylbenzene
SMILES CCCCCCCCCCCCCCCCCCC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 32°C to 36°C
Color White to Yellow
Boiling Point 249°C (15.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5(CH2)17CH3
Quantity 50 g
Synonym 1-naphthalen-2-yl prop-2-en-1-one, 1-2-naphthyl prop-2-en-1-one, jak3 inhibitor v, 2-naphthylvinyl ketone, 2'-acrylonaphthone, 2-acryloylnaphthalene, thr1 inhibitor, 1, 2-naphthyl vinyl ketone, biomolki_000064, biomolki2_000005
PubChem CID 3799
Formula Weight 330.59
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material Octadecylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.