missing translation for 'onlineSavingsMsg'
Learn More

Octadecyltrimethoxysilane, 90%, Tech.

Product Code. 10657251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10657251 25 mL Glass bottle 25mL
10300994 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10657251 Supplier Thermo Scientific Acros Supplier No. 313600250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3069-42-9 | C21H46O3Si | 374.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3069-42-9
Molecular Formula C21H46O3Si
Molecular Weight (g/mol) 374.69
MDL Number MFCD00043060
InChI Key SLYCYWCVSGPDFR-UHFFFAOYSA-N
Synonym octadecyltrimethoxysilane, trimethoxy octadecyl silane, n-octadecyltrimethoxysilane, stearyltrimethoxysilane, silane, trimethoxyoctadecyl, octadecyl-trimethoxysilane, acmc-209hi7, ksc491g4l, trimethoxy octadecyl silane, technical grade
PubChem CID 76486
IUPAC Name trimethoxy(octadecyl)silane
SMILES CCCCCCCCCCCCCCCCCC[Si](OC)(OC)OC

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 13°C to 17°C
Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 170°C (0.1 mmHg)
Flash Point 139°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. Total octadecyltrimethoxysilanes (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)17Si(OCH3)3
Refractive Index 1.4380 to 1.44
Quantity 25 mL
Specific Gravity 0.88
Solubility Information Solubility in water: insoluble (reacts)
Formula Weight 374.69
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Octadecyltrimethoxysilane
Show More Show Less
compliance-icons
Product Identifier
  • Octadecyltrimethoxysilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Octadecyltrimethoxysilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.