Learn More
Fisherbrand™ Octahedral PTFE Stir Bar
Provides stable stirring action. These PTFE magnetic stir bars are autoclavable and include a pivot ring. The flat surfaces provide a great mixing action.
Brand:  Fisherbrand™
Code : 41
Additional Details : Weight : 0.05580kg
 View more versions of this product
Product Code. 11527782
  • 19.30 € / Pack of 10
In Stock
to see stock.
Add to basket
Description
Description
  • Stable, general-purpose magnetic stir bar
  • The flat surfaces provide a greater mixing action
  • All have pivot rings
  • AlNiCo-V magnets encased in PTFE
  • Autoclavable
Specifications
Specifications
9.5 mm
25 x 9.5mm
10/Pk.
25 mm