Learn More
Fisherbrand™ Octahedral PTFE Stir Bar
Provides stable stirring action. These PTFE magnetic stir bars are autoclavable and include a pivot ring. The flat surfaces provide a great mixing action.
9.70 € - 55.35 €
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
11577772
|
Fisherbrand™
|
13 mm | 8 mm |
9.70 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11507782
|
Fisherbrand™
|
38 mm | 8 mm |
21.95 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11537782
|
Fisherbrand™
|
38 mm | 9.5 mm |
20.25 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11547782
|
Fisherbrand™
|
51 mm | 9.5 mm |
44.05 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11527782
|
Fisherbrand™
|
25 mm | 9.5 mm |
19.30 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11557782
|
Fisherbrand™
|
64 mm | 9.5 mm |
55.35 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
Description
- Stable, general-purpose magnetic stir bar
- The flat surfaces provide a greater mixing action
- All have pivot rings
- AlNiCo-V magnets encased in PTFE
- Autoclavable
Specifications
10/Pk. |