missing translation for 'onlineSavingsMsg'
Learn More

Octan-1-ol, Pure, Fisher Chemical™

Product Code. 10121960 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
Unit Size:
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10121960 2.5 L Amber glass bottle 2.5 L
10306290 500 mL Amber glass bottle 500mL
11448443 2.5 L Plastic Coated Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10121960 Supplier Fisher Chemical Supplier No. O/0050/17
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Specifications

Melting Point -16°C
Color Colorless
Boiling Point 195°C
Packaging Amber glass bottle
Quantity 2.5 L
Vapor Density 4.5
Formula Weight 130.23g/mol
Physical Form Liquid
compliance-icons
Product Identifier
  • 1-Octanol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.