missing translation for 'onlineSavingsMsg'
Learn More

Octan-2-ol, 98+%, Pure, Fisher Chemical™

Product Code. 10627612 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10627612 2.5 L 2.5 L
10598620 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10627612 Supplier Fisher Chemical Supplier No. O/0100/17
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

C8H18O, CAS Number-123-96-6, sec-caprylic alcohol, n-octan-2-ol, methylhexylcarbinol, hexylmethylcarbinol, 2-octyl alcohol, 2-hydroxyoctane, dl-2-octanol, 2-octanol, 1-methylheptyl alcohol, 1-methylheptanol, 2.5L, 174 deg.C, CHEBI:37869, Yellow, 130.23g/mol, SJWFXCIHNDVPSH-UHFFFAOYSA-N, -38 deg.C

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 123-96-6
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number 4591
InChI Key SJWFXCIHNDVPSH-UHFFFAOYSA-N
Synonym 2-octanol, 2-octyl alcohol, hexylmethylcarbinol, 2-hydroxyoctane, sec-caprylic alcohol, 1-methylheptanol, methylhexylcarbinol, 1-methylheptyl alcohol, dl-2-octanol, n-octan-2-ol
PubChem CID 20083
ChEBI CHEBI:37869
IUPAC Name octan-2-ol
SMILES CCCCCCC(C)O

Specifikationer

Melting Point -38°C
Color Yellow
Boiling Point 174°C
Packaging Amber glass bottle
Quantity 2.5 L
Formula Weight 130.23g/mol
Physical Form Liquid
compliance-icons
Produktbeteckning
  • DL-2-Octanol
Signalord
  • Danger
Riskkategorier
  • Serious eye damage/eye irritation Category 1
Faroangivelser
  • H318-Causes serious eye damage.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.