missing translation for 'onlineSavingsMsg'
Learn More

Octanoyl chloride, 99%

Product Code. 10607292 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10607292 100 g Glass bottle 100g
10697092 5 g Glass bottle 5g
10489050 500 g Glass bottle 500g
10249583 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10607292 Supplier Thermo Scientific Acros Supplier No. 129431000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 111-64-8 | C8H15ClO | 162.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-64-8
MDL Number MFCD00000766
InChI Key REEZZSHJLXOIHL-UHFFFAOYSA-N
Synonym capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride
PubChem CID 8124
IUPAC Name octanoyl chloride
SMILES CCCCCCCC(=O)Cl

Specifications

Acidity 1% max.
CAS Min % 98.5
CAS Max % 100.0
Melting Point -63.0°C
Density 0.9500g/mL
Boiling Point 195.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H15ClO
Linear Formula CH3(CH2)6COCl
Refractive Index 1.4340 to 1.4360
Quantity 100 g
Beilstein 02, 348
Specific Gravity 0.95
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohol, miscible with most common organic solvents, soluble in ether
Molecular Weight (g/mol) 162.66
Viscosity 1.51 mPa.s (20°C)
Formula Weight 162.66
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Octanoyl chloride, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Octanoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.