missing translation for 'onlineSavingsMsg'
Learn More

Octanoyl Chloride, TRC

Product Code. 30426460
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426460 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426460 Supplier Toronto Research Chemicals Supplier No. TRCO2397401G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octanoyl Chloride
CAS 111-64-8
Molecular Formula C8H15ClO
Quantity 1 g
Synonym 1-Octanoic Acid Chloride, Caprylic Acid Chloride, Capryloyl Chloride, Caprylyl Chloride, Octanoic Acid Chloride, Octanoic Chloride, n-Octanoyl Chloride
InChI Formula InChI=1S/C8H15ClO/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3
SMILES CCCCCCCC(=O)Cl
IUPAC Name octanoyl chloride
Molecular Weight (g/mol) 162.66
Formula Weight 162.0811
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.