missing translation for 'onlineSavingsMsg'
Learn More

Octazamide, MedChemExpress

Product Code. 16453149
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453149 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16453149 Supplier Medchem Express Supplier No. HYU00101
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Octazamide (ICI-US 457) is an analgesic drug.

Octazamide (ICI-US 457) is an analgesic drug.

Specifications

Chemical Name or Material Octazamide
CAS 56391-55-0
Color Colorless
Purity Grade Notes Research
Molecular Formula C13H15NO2
Quantity 1 mg
Synonym ICI-US 457
SMILES O=C(C1=CC=CC=C1)N2CC(COC3)C3C2
Molecular Weight (g/mol) 217.26
Formula Weight 217.26
Percent Purity 98.55%
Grade Research
Physical Form Liquid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.