missing translation for 'onlineSavingsMsg'
Learn More

Octenidine dihydrochloride, 98%

Product Code. 15438808
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438808 5 g 5g
15428808 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438808 Supplier Thermo Scientific Alfa Aesar Supplier No. H62257.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 70775-75-6 | C36H64Cl2N4

Octenidine dihydrochloride is used as a antiseptic for skin, mucous Membranes and wounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octenidine dihydrochloride is used as a antiseptic for skin, mucous Membranes and wounds.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 70775-75-6
Molecular Formula C36H64Cl2N4
MDL Number MFCD01938808
Synonym N,N'-[Decane-1,10-diyldi-1(4H)-pyridyl-4-ylidene]bis(octylammonium) dichloride

Especificaciones

Melting Point 214°C
Quantity 5 g
Merck Index 14,6754
Solubility Information Soluble in dimethyl sulfoxide and methanol.
Formula Weight 623.83
Percent Purity 98%
Chemical Name or Material Octenidine dihydrochloride
compliance-icons
Nombre del producto
  • Octenidine dihydrochloride
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contain: N,N'-(Decane-1,10-diyldi-1(4H)-pyridyl-4-yli dene)bis(octylammonium) dichloride

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.