missing translation for 'onlineSavingsMsg'
Learn More

Octenidine Dihydrochloride, TRC

Product Code. 17973805
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973805 25 g 25g
17922835 1 g 1g
17910345 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17973805 Supplier Toronto Research Chemicals Supplier No. TRCO23915025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70775-75-6
Molecular Formula C36 H62 N4 . 2 H Cl
Molecular Weight (g/mol) 623.83
Synonym N,N′-(1,10-decanediyldi-1(4H)-pyridinyl-4-ylidene)bis-1-Octanamine Dihydrochloride, Neo Kodan, Octeniderm, Octenidine 2HCl;Octenidine Dihydrochloride, Octenidine Hydrochloride, Octenidol, Octenilin, Win 41464-2
IUPAC Name N-octyl-1-[10-[4-(octylamino)pyridin-1-ium-1-yl]decyl]pyridin-1-ium-4-amine;dichloride
SMILES CCCCCCCC/N=C(C=C1)/C=CN1CCCCCCCCCCN(C=C/2)C=CC2=N/CCCCCCCC.[2ClH]

Specifications

Quantity 25 g
InChI Formula InChI=1S/C36H62N4.2ClH/c1-3-5-7-9-15-19-27-37-35-23-31-39(32-24-35)29-21-17-13-11-12-14-18-22-30-40-33-25-36(26-34-40)38-28-20-16-10-8-6-4-2;;/h23-26,31-34H,3-22,27-30H2,1-2H3;2*1H
Formula Weight 622.4508
Percent Purity >95%
Chemical Name or Material Octenidine Dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.