missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octocrylene, MedChemExpress
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Description
Octocrylene is an organic ultraviolet (UV) filter which absorbs mainly UVB radiation and short UVA wavelengths. Octocrylene has the potential for using in various cosmetic products to either provide an appropriate sun protection factor in sunscreen products or to protect cosmetic formulations from UV radiation.
Specifications
Specifications
| Chemical Name or Material | Octocrylene |
| CAS | 6197-30-4 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C24H27NO2 |
| Quantity | 5 g |
| Solubility Information | DMSO : ≥ 100 mg/mL (276.64 mM) ∣Ethanol : 50 mg/mL (138.32 mM; Need ultrasonic) |
| SMILES | O=C(OCC(CC)CCCC)/C(C#N)=C(C1=CC=CC=C1)\C2=CC=CC=C2 |
| Molecular Weight (g/mol) | 361.48 |
| Formula Weight | 361.48 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction