missing translation for 'onlineSavingsMsg'
Learn More

Octyl beta-D-galactopyranoside, TRC

Product Code. 30008964
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008964 250 mg 250mg
30005209 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008964 Supplier Toronto Research Chemicals Supplier No. TRCO295003250MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40427-75-6
Molecular Formula C14H28O6
Molecular Weight (g/mol) 292.37
Synonym β-D-Galactopyranoside, octyl (9CI, ACI), Galactopyranoside, octyl, β-D- (6CI), Octyl β-D-galactopyranoside (ACI), 1-O-Octyl-β-D-galactopyranoside, Octyl β-galactoside
IUPAC Name (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol
SMILES O[C@@H]([C@@H](CO)O[C@@H](OCCCCCCCC)[C@@H]1O)[C@@H]1O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-13(18)12(17)11(16)10(9-15)20-14/h10-18H,2-9H2,1H3/t10-,11+,12+,13-,14-/m1/s1
Formula Weight 292.1886
Chemical Name or Material Octyl beta-D-galactopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.