missing translation for 'onlineSavingsMsg'
Learn More

Octyl beta-D-Glucopyranoside, TRC

Product Code. 30137443
Change view
Click to view available options
Quantity:
100 mg
5 g
10 g
Unit Size:
100mg
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137443 10 g 10g
30134697 5 g 5g
30009689 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137443 Supplier Toronto Research Chemicals Supplier No. TRCO28600010G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29836-26-8
Molecular Formula C14 H28 O6
Molecular Weight (g/mol) 292.37
Synonym β-D-Glucopyranoside, octyl (9CI, ACI), Glucopyranoside, octyl, β-D- (8CI), Octyl β-D-glucopyranoside (ACI), 1-n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, 1-O-Octyl-β-D-glucopyranose, 1-O-Octyl-β-D-glucopyranoside, 1-Octyl β-D-glucopyranoside, 1-Octyl β-D-glucoside, BTB 11967, n-Octyl β-D-glucopyranoside, n-Octyl β-D-glucoside, Octyl glucoside, Octyl β-D-glucoside, Octyl β-D-glycopyranoside, Octyl β-glucopyranoside, PlantaCare 810, β-D-Octyl glucoside, β-Octyl monoglucoside
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-13(18)12(17)11(16)10(9-15)20-14/h10-18H,2-9H2,1H3/t10-,11-,12+,13-,14-/m1/s1
Formula Weight 292.1886
Chemical Name or Material N-Octyl-beta-D-glucopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.