missing translation for 'onlineSavingsMsg'
Learn More

Octyl isocyanate, 99%

Product Code. 10686504 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10686504 5 g Glass bottle 5g
10516992 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10686504 Supplier Thermo Scientific Acros Supplier No. 416460050
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3158-26-7 | C9H17NO | 155.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3158-26-7
Molecular Formula C9H17NO
Molecular Weight (g/mol) 155.24
MDL Number MFCD00013887
InChI Key DYQFCTCUULUMTQ-UHFFFAOYSA-N
Synonym octyl isocyanate, octylisocyanate, octane, 1-isocyanato, n-octylisocyanate, 1-octyl isocyanate, n-octyl isocyanate, 1-isocyanato-octane, isocyanic acid, octyl ester, octanisocyanate
PubChem CID 76616
IUPAC Name 1-isocyanatooctane
SMILES CCCCCCCCN=C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Boiling Point 200°C to 204°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7NCO
Refractive Index 1.4290 to 1.431
Quantity 5 g
Solubility Information Solubility in water: decomposes in water
Formula Weight 155.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Octyl isocyanate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Octyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.