missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Octyl Sepharose™ 4 Fast Flow Chromatography Media

Product Code. 10039649 Shop All Cytiva Products
Click to view available options
Quantity:
25 mL
200 mL
Unit Size:
25mL
Each
This item is not returnable. View return policy

Product Code. 10039649

Brand: Cytiva 17094602

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Aliphatic hydrophobic interaction chromatography (HIC) medium for capture and intermediate purification of larger proteins

  • HIC ligands are coupled to monosacharide units via their corresponding glycidylethers, giving matrices without charges and stable etherbonds between ligands and agarose
  • Octyl derivative adds different and complementary selectivity to Sepharose Fast Flow HIC product family
  • Resin based on cross-linked 4% agarose
  • Supported for industrial applications and well established in approved processes
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 mL
Stability Stable in commonly used aqueous Buffers: 1 M NaOH, 30% Isopropanol, 70% Ethanol, 6 M Guanidine-Hydrochloride, 30% Acetonitrile, 1 mM HCl
For Use With (Application) For Capture and Intermediate PurifiCatIon of Larger Proteins
Storage Requirements 4°C to 30°C
Product Type Octyl Sepharose 4 Fast Flow Media
Flow Rate 150 to 250 cm/Hoμr
Particle Size 45 to 165 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Max. Pressure 100 kPa
Matrix 4% Cross-Linked Agarose
pH Range 2 to 14
Storage Buffer 20% Ethanol
Ligand Type Octyl
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.