Learn More
Cytiva Octyl Sepharose™ 4 Fast Flow Chromatography Media
Aliphatic hydrophobic interaction chromatography (HIC) medium for capture and intermediate purification of larger proteins
Brand: Cytiva 17-0946-10
Code : GE
Additional Details : CAS Number : 64-17-5 Weight : 0.11000kg
Description
- HIC ligands are coupled to monosacharide units via their corresponding glycidylethers, giving matrices without charges and stable etherbonds between ligands and agarose
- Octyl derivative adds different and complementary selectivity to Sepharose Fast Flow HIC product family
- Resin based on cross-linked 4% agarose
- Supported for industrial applications and well established in approved processes
- Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
Specifications
25 mL | |
Stable in commonly used aqueous Buffers: 1 M NaOH, 30% Isopropanol, 70% Ethanol, 6 M Guanidine-Hydrochloride, 30% Acetonitrile, 1 mM HCl | |
Octyl Sepharose 4 Fast Flow Media | |
Resin on cross-linked 4% agarose | |
4% Cross-Linked Agarose | |
20% Ethanol |
For Capture/Intermediate Purification of Large Proteins | |
4°C to 30°C | |
45 to 165 μm | |
100 kPa | |
2 to 14 | |
Octyl |
Safety and Handling
- Octyl Sepharose 4 Fast Flow, 25 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol