missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Octyl Sepharose™ 4 Fast Flow Chromatography Media

Product Code. p-4592264 Shop All Cytiva Products
Click to view available options
Quantity:
25 mL
200 mL
Unit Size:
25mL
Each
This item is not returnable. View return policy

Product Code. 10049649

Brand: Cytiva 17094610

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Aliphatic hydrophobic interaction chromatography (HIC) medium for capture and intermediate purification of larger proteins

  • HIC ligands are coupled to monosacharide units via their corresponding glycidylethers, giving matrices without charges and stable etherbonds between ligands and agarose
  • Octyl derivative adds different and complementary selectivity to Sepharose Fast Flow HIC product family
  • Resin based on cross-linked 4% agarose
  • Supported for industrial applications and well established in approved processes
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mL
Stability Stable in commonly used aqueous Buffers: 1 M NaOH, 30% Isopropanol, 70% Ethanol, 6 M Guanidine-Hydrochloride, 30% Acetonitrile, 1 mM HCl
For Use With (Application) For Capture/Intermediate Purification of Large Proteins
Storage Requirements 4°C to 30°C
Product Type Octyl Sepharose 4 Fast Flow Media
Particle Size 45 to 165 μm
Material Resin on cross-linked 4% agarose
Max. Pressure 100 kPa
Matrix 4% Cross-Linked Agarose
pH Range 2 to 14
Storage Buffer 20% Ethanol
Ligand Type Octyl
Show More Show Less
compliance-icons
Product Identifier
  • Octyl Sepharose 4 Fast Flow, 25 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.