missing translation for 'onlineSavingsMsg'
Learn More

Octyl sulfate, sodium salt, 99%, HPLC grade

Product Code. 10576602 Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10576602

Brand: Thermo Scientific 416500250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142-31-4 | C8H15Na·H2SO4 | 232.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ion pair chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-31-4
Molecular Formula C8H15Na·H2SO4
Molecular Weight (g/mol) 232.28
InChI Key WFRKJMRGXGWHBM-UHFFFAOYSA-M
Synonym sodium octyl sulfate, sodium n-octyl sulfate, sodium octyl sulphate, sipex ols, cycloryl os, duponol 80, sodium capryl sulfate, octyl sodium sulfate, sulfuric acid, monooctyl ester, sodium salt, octyl sulfate sodium salt
PubChem CID 2735107
IUPAC Name sodium;octyl sulfate
SMILES CCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Absorbance (5% aq. soln.) (1cm cell), (5% aq. soln.) (1 cm cell) at 0nm, 0.02 max. at 250nm
Melting Point 210.0°C
Color White
Loss on Drying 2% (105°C) (vacuum/P2O5) max.
Infrared Spectrum Authentic
Assay Percent Range 1% max. Alkyl sulphate homologues (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7OSO3Na
Quantity 25 g
Solubility Information (25% in water at 25°C) Clear colorless
Formula Weight 232.28
Percent Purity 99%
Physical Form Coarse Powder
Chemical Name or Material Octyl sulfate, sodium salt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.