missing translation for 'onlineSavingsMsg'
Learn More

Octyl sulfate, sodium salt, 99%, HPLC grade
GREENER_CHOICE

Product Code. 11985151 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11985151 100 g Glass bottle 100g
10576602 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11985151 Supplier Thermo Scientific Supplier No. 416501000
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 142-31-4 | C8H15Na·H2SO4 | 232.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ion pair chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-31-4
Molecular Formula C8H15Na·H2SO4
Molecular Weight (g/mol) 232.28
InChI Key WFRKJMRGXGWHBM-UHFFFAOYSA-M
Synonym sodium octyl sulfate, sodium n-octyl sulfate, sodium octyl sulphate, sipex ols, cycloryl os, duponol 80, sodium capryl sulfate, octyl sodium sulfate, sulfuric acid, monooctyl ester, sodium salt, octyl sulfate sodium salt
PubChem CID 2735107
IUPAC Name sodium;octyl sulfate
SMILES CCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Absorbance (5% aq. soln.) (1cm cell), (5% aq. soln.) (1 cm cell) at 0nm, 0.02 max. at 250nm
Melting Point 210.0°C
Color White
Loss on Drying 2% (105°C) (vacuum/P2O5) max.
Infrared Spectrum Authentic
Assay Percent Range 1% max. Alkyl sulphate homologues (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7OSO3Na
Quantity 100 g
Solubility Information (25% in water at 25°C) Clear colorless
Formula Weight 232.28
Percent Purity 99%
Physical Form Coarse Powder
Chemical Name or Material Octyl sulfate, sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.