missing translation for 'onlineSavingsMsg'
Learn More

Octylamine, TRC

Product Code. 30426879
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426879 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426879 Supplier Toronto Research Chemicals Supplier No. TRCO270815250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octylamine
CAS 111-86-4
Molecular Formula C8 H19 N
Quantity 250 g
Synonym 1-Octanamine (9CI','ACI), Octylamine (8CI), 1-Aminooctane, 1-Octylamine, Amine OD, Armeen 8, Armeen 8D, Caprylylamine, Farmin 08D, Genamin 8R, Lipomin 8D, Monooctylamine, n-Octylamine, NSC 9824, Octan-1-amine, Octanamine
InChI Formula InChI=1S/C8H19N/c1-2-3-4-5-6-7-8-9/h2-9H2,1H3
SMILES CCCCCCCCN
IUPAC Name octan-1-amine
Molecular Weight (g/mol) 129.24
Formula Weight 129.1517
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.