missing translation for 'onlineSavingsMsg'
Learn More

Octyltrichlorosilane, 97%

Product Code. 10388490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10388490 100 g 100g
10269273 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10388490 Supplier Thermo Scientific Acros Supplier No. 216501000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5283-66-9 | C8H17Cl3Si | 247.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5283-66-9
MDL Number MFCD00000488
InChI Key RCHUVCPBWWSUMC-UHFFFAOYSA-N
Synonym trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive
PubChem CID 21354
IUPAC Name trichloro(octyl)silane
SMILES CCCCCCCC[Si](Cl)(Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 233°C (731.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H17Cl3Si
Linear Formula CH3(CH2)7SiCl3
Refractive Index 1.4470 to 1.449
Quantity 100 g
Beilstein 04, III, 1907
Specific Gravity 1.07
Solubility Information Solubility in water: reacts
Molecular Weight (g/mol) 247.67
Viscosity 1.9 mPa.s (25°C)
Formula Weight 247.67
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Octyltrichlorosilane
Show More Show Less
compliance-icons
Product Identifier
  • Octyltrichlorosilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/physician.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.