missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ ODQ

Product Code. 30001137
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001137 50 mg 50mg
30001136 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001137 Supplier R&D Systems™ Supplier No. 0880/50
  • ONLINE EXCLUSIVE PRICE
    673.21 €
    606.90 € / 50mg
    Save 66.31 €
    9% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of NO-sensitive guanylyl cyclase

A potent and selective inhibitor of NO-sensitive guanylyl cyclase.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41443-28-1
Molecular Formula C9H5N3O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 187.16
Purity >99%
Molecular Weight (g/mol) 187.158
Chemical Name or Material ODQ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.