missing translation for 'onlineSavingsMsg'
Learn More

Officinalisinin I, MedChemExpress

Product Code. 16406817
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16406817 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16406817 Supplier Medchem Express Supplier No. HY107284
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Officinalisinin I is a steroidal saponin, isolated from Anemarrhena asphodeloides.

Officinalisinin I is a steroidal saponin, isolated from Anemarrhena asphodeloides.

Specifications

Chemical Name or Material Officinalisinin I
CAS 57944-18-0
Color White
Purity Grade Notes Research
Molecular Formula C45H76O19
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (108.57 mM; Need ultrasonic)
SMILES C[C@@]12[C@](C[C@@]3([H])[C@]2([H])[C@@H]([C@](CCC(C)CO[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)CO)(O)O3)C)([H])[C@@]5([H])[C@]([C@@]6([C@](C[C@@H](O[C@@]7([H])[C@@H]([C@H]([C@H](O)[C@@H](CO)O7)O)O[C@]8([H])O[C@@H]([C@@H](O)[C@H](O)[C@H]8O)CO)CC6)([H])CC5)C)([H])CC1
Molecular Weight (g/mol) 921.07
Formula Weight 921.07
Percent Purity 98.99%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.