missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OG-L002

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
OG-L002 is a potent LSD1 inhibitor (IC50 = 20 nM). Exhibits some selectivity for LSD1 over MAO-B and MAO-A (IC50 values are 0.72 and 1.38 μM, respectively). Inhibits herpes simplex virus (HSV) immediate early gene expression and viral yield in vitro. Suppresses HSV primary infection in vivo.
Specifications
Specifications
| Chemical Name or Material | 4 '-((1 R,2 S)-2-aminocyclopropyl)biphenyl-3-ol |
| CAS | 1357302-64-7 |
| Synonym | 4′-((1R,2S)-2-aminocyclopropyl)biphenyl-3-ol |
| Quantity | 5 mg |
| Target | Histone Demethylase Inhibitors |
| Molecular Formula | C15H15NO |
| Formula Weight | 225.29 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction