Learn More
OG-L002
Potent LSD1 inhibitor
Brand: Tocris Bioscience 6244/25
This item is not returnable.
View return policy
Description
OG-L002 is a potent LSD1 inhibitor (IC50 = 20 nM). Exhibits some selectivity for LSD1 over MAO-B and MAO-A (IC50 values are 0.72 and 1.38 μM, respectively). Inhibits herpes simplex virus (HSV) immediate early gene expression and viral yield in vitro. Suppresses HSV primary infection in vivo.Specifications
| 4 '-((1 R,2 S)-2-aminocyclopropyl)biphenyl-3-ol | |
| 1357302-64-7 | |
| 4′-((1R,2S)-2-aminocyclopropyl)biphenyl-3-ol | |
| 25 mg |