Learn More
Tocris Bioscience™ Ogerin
GREENER_CHOICE
Selective positive allosteric modulator at GPR68
Brand:  Tocris Bioscience™ 5722/10
Encompass
Product Code. 15737076
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Ogerin is a selective positive allosteric modulator (PAM) of GPR68 (pEC50 = 6.83), which exhibits selectivity for GPR68 over 58 other GPCRs. Ogerin attenuates contextual-based fear memory in mice.
Specifications
Specifications
2-[4-Amino-6-[(phenylmethyl)amino]-1,3,5-triazin-2-yl]benzenemethanol
1309198-71-7