missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Adventurer™ Precision Balance

Product Code. 15501936 Shop All Ohaus Products
Click to view available options
Model:
AX4201
AX422
AX8201
AX822
Capacity:
420 g
820 g
4200 g
8200 g
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15501936

Brand: OHAUS™ 30206081

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Adventurer™ Precision Balance strikes the ideal balance between inventive features and functional, uncomplicated weighing capabilities.

Delivers stable and reliable weighing results for routine weighing tasks.

  • Fast stabilization time improves productivity in the laboratory
  • AutoCal™ OHAUS automatic internal calibration system performs routine maintenance by calibrating the balance daily
  • Color touchscreen offers easy and fast operation of Adventurer applications
  • Operate and access nine application modes and abundant features that eliminate the need to do several manual calculations through the modern color touchscreen
  • Operators can wear laboratory gloves while utilizing the touchscreen, eliminating the inconvenience and hazards associated with constantly putting on and removing gloves
  • In addition to the touchscreen, Adventurer also has six mechanical keys that provide tactile feedback and allow the operator to perform repetitive operations such as tare, zero, calibration, and print
  • Application modes: weighing, dynamic weighing, display hold, parts counting, density determination, totalization, percent weighing, check weighing, formulation
  • Real-time clock function keeps accurate time even during power loss and the GLP data capability has the ability to record Sample name, Project names and Balance IDs to help meet traceability and compliance requirements
  • Cloning feature allows you to save user and application settings to a USB flash drive which can be easily used to configure additional Adventurer balances
  • When using the minimum weight feature, the display clearly indicates that your current sample weight is below your defined minimum limit; simply increase your sample weight to assure that your results are up to your standards
  • Some models have a space-saving draftshield designed to improve user experience and accessibility.
  • Additional features include: RS232 interface, integral weigh below hook, full housing in-use cover, removable stainless steel pan, die-cast metal bottom housing, security bracket, illuminated up-front level indicator, four adjustable feet, software lockout menus, stability indicator, software overload/underload indicators, user selectable environmental settings, audible indicator, user selectable brightness settings, auto dim, touchscreen calibration, auto tare, user selectable operating language (nine)

Dual USB Ports

  • A front USB host port is easily accessible and makes it simple to load data from the balance on to a flash drive without having to reach around to the back or move the balance
  • A second USB device is located at the rear of the balance that can be used to connect the balance to a PC
  • The connectivity options help meet traceability requirements in traditional installations

Compliance

Metrology: OIML R76, EN 45501 (Class I, nmax 320000, Class II, nmax 52000), Product Safety: EN 61010-1, IEC 61010-1, Electromagnetic Compatibility: IEC 61326-1, EN 61326-1 (Emissions Class B, Immunity Industrial Requirements), GLP/GMP Compliant

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Metrology: OIML R76, EN 45501 (Class I, nmax 320000, Class II, nmax 52000), Product Safety: EN 61010-1, IEC 61010-1, Electromagnetic Compatibility: IEC 61326-1, EN 61326-1 (Emissions Class B, Immunity Industrial Requirements), GLP/GMP Compliant
Connection Type RS232, USB Host, USB Device
Description Precision Balance AX822
Capacity 820 g
Display Type Color Touchscreen
Readability 0.01 g
For Use With (Application) Weighing, Parts Counting, Percent Weighing, Check
Includes AC Adapter
Linearity ±0.02 g
Material Metal Base, ABS Top Housing, Stainless Steel Pan
Measurement Time 1.5 sec.
Min. Sample Weight 8.2 g
Model AX822
Repeatability 0.01 g
Sensitivity Drift 3 ppm/°C
Stabilization Time 1.5 sec.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C
Test Load 800 g, OIML F1 or ASTM CL 2
Type Toploading Balances
Warranty 2 years
Weighing Modes Weighing (17 units + custom unit), parts counting, percentage weighing, check weighing, animal/dynamic weighing, totalization/statistics, formulation, density determination, display hold, batch printing
Dimensions (L x W x H) 354 x 230 x 100 mm
Length (English) 13.93 in.
Length (Metric) 354 mm
Width (English) Pan 6.9 in.
Width (Metric) Pan 175 mm
Height (Metric) 100 mm
Height (Metric) Pan 195 mm
Depth (English) Pan 7.7 in.
Depth (Metric) 354 mm
Depth (Metric) Pan 195 mm
Weight (English) 14.3 lb.
Weight (Metric) 6.5 kg
Electrical Requirements AC Adapter Input: 100/240 V, 0.3 A, 50/60 Hz
Voltage 100/240 V
Amperage AC Adapter Input: 0.3 A, AC Adapter Output: 0.84 A
Frequency AC Adapter Input: 50/60 Hz
Legal for Trade No
Calibration Internal
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.