Learn More
OHAUS™ Interface Kit for Navigator, Scout Pro, and TA Series Balances
Product Code. 11752158 Shop All Ohaus Products
Change view
Click to view available options
Connection Type:
RS-232
USB
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connection Type Unit Size
11752158 USB Each
11742158 RS-232 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11752158 Supplier OHAUS™ Supplier No. 83032108
  • ONLINE EXCLUSIVE PRICE
    51.00 €
    38.41 € / Pack of 1
    Save 12.59 €
    24% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Connects Navigator Series balances to peripherals, including printers and monitors.
  • For use with Navigator, Scout Pro, and TA Series Balances
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) NV/NVL/NVT/SP/TA Balance
Connection Type USB
Type USB Accessory Kit