Learn More
Draeger Safety Oil Impactor
Product Code. 15644994
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. Quantity Unit Size
15644994 10/Pk. Pack of 10
1 options
Product Code. 15644994 Supplier Draeger Safety Supplier No. 8103530
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Draeger Safety Oil Impactor tests for all synthetic and mineral oil mists in compressed-air and breathing-air.
  • Utilizing an innovative oil detection process the impactor eliminates the need to use an oxidizing reaction to detect for the presence of oil
Specifications
Description Wide
Quantity 10/Pk.