Learn More
Justrite™ Oily Waste Can Countertop
Product Code. 12913565
Product Code. 12913565 Supplier Justrite™ Supplier No. 09200Y
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Lid opens no more than 60 degrees, steel, powder coated, for disposal of cloths, rags, wipers, soaked in oil, solvents yellow 8L