Learn More
Ohaus™ OIML Class E2 Weights
Shop All Ohaus Products
Click to view available options
:
Weight: 0.030 kg
Description
- Constructed of stainless steel
- Weights include a weight case
- Single body design without adjusting cavity
Specifications
Specifications
| Classification | OIML E2 Weights |
| Weight (Metric) | 0.030 kg |