missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ OIML Class F1 Proof Line Weight Set without Certificate

Product Code. 11363354 Shop All Sartorius Products
Change view
Click to view available options
Mass:
1 g to 1 kg
1 g to 100 g
1 g to 5 kg
1 mg to 500 mg
Material:
Aluminum (1 to 5 mg), Nickel Silver (10 to 500 mg)
Special Stainless Steel
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mass Material Unit Size
11363354 1 mg to 500 mg Aluminum (1 to 5 mg), Nickel Silver (10 to 500 mg) Each
11333354 1 g to 100 g Special Stainless Steel Each
11323374 1 g to 1 kg Special Stainless Steel Each
11303384 1 g to 5 kg Special Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11363354 Supplier Sartorius™ Supplier No. YCS01253AC00
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Includes the same features and properties as the individual weights of this series, along with the corresponding maximum permissible errors. Sartorius™ OIML Class F1 Proof LIne Weight Set without Certificate is supplied with gloves, tweezers and brush in high-quality plastic case

Values 1 to 5mg:
Aluminum leaf weights with density of 2,650 kg/m3

Values of 10 to 500mg:
Nickel silver construction with density of 8,600 kg/m3.

Values of 1g and higher:
Polished, special stainless steel knob weights with density of 7,950kg/m3

  • Highly corrosion resistant
  • Non-magnetizable
  • Tolerances comply with OIML R111-1
  • Includes sturdy plastic carry case
TRUSTED_SUSTAINABILITY

Specifications

Product Type Proof Line Weight Set
Classification OIML Class F1
Material Aluminum (1 to 5 mg), Nickel Silver (10 to 500 mg)
Mass 1 mg to 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.