missing translation for 'onlineSavingsMsg'
Learn More

Olanzapine (1mg/ml in Acetonitrile), TRC

Product Code. 30400842
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400842 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400842 Supplier Toronto Research Chemicals Supplier No. TRCKIT05905X1ML
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 132539-06-1
Molecular Formula C17 H20 N4 S
Quantity 5 x 1 mL
Synonym 10H-Thieno[2,3-b][1,5]benzodiazepine, 2-methyl-4-(4-methyl-1-piperazinyl)-, 2-Methyl-4-(4-methyl-1-piperazinyl)-10H-thieno[2,3-b][1,5]benzodiazepine, ADAMED, Arenbil, Integrol, LY 170053, Lanzac, Oferta, Olanz, Olanzapine, Oleanz, Olenz, Olimelt, Oliza, Oltal, Prexal, Rexapin, Zolafren, Zolafren Flas, ZypAdhera, Zyprexa, Zyprexa-Velotab, Zyprobiox
InChI Formula InChI=1S/C17H20N4S/c1-12-11-13-16(21-9-7-20(2)8-10-21)18-14-5-3-4-6-15(14)19-17(13)22-12/h3-6,11,19H,7-10H2,1-2H3
SMILES CN1CCN(CC1)C2=Nc3ccccc3Nc4sc(C)cc24
IUPAC Name 2-methyl-4-(4-methylpiperazin-1-yl)-10H-thieno[2,3-b][1,5]benzodiazepine
Molecular Weight (g/mol) 312.43
Formula Weight 312.1409
Concentration or Composition (by Analyte or Components) Olanzapine 1 mg/mL
Chemical Name or Material Olanzapine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.