missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Olaparib
GREENER_CHOICE

Product Code. 18719856
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18719856 10 mg 10mg
18799536 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18719856 Supplier Tocris Bioscience™ Supplier No. 7579/10
  • ONLINE EXCLUSIVE PRICE
    113.00 €
    106.05 € / 10mg
    Save 6.95 €
    6% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent PARP inhibitor

Olaparib is a potent PARP2 and PARP1 inhibitor (IC50 values are 1 and 5 nM, respectively). Olaparib abolishes PARP-1 activity at a concentration of 30-100 nM in SW620 cell lysates. BRCA1-deficient cell lines are hypersensitive to PARP inhibition by Olaparib in comparison with BRCA1- and BRCA2-proficient cells. Olaparib potentiates the antitumor activity of the methylating chemotherapeutic agent Temozolomide (Cat. No. 2706) in xenograft mouse model. Olaparib is orally bioavailable in rats.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[[3-[[4-(Cyclopropylcarbonyl)-1-piperazinyl]carbonyl]-4-fluorophenyl]methyl]-1(2 H)-phthalazinone
CAS 763113-22-0
Quantity 10 mg
Target Poly(ADP-ribose) Polymerase Inhibitors
Molecular Formula C24H23FN4O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.