missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Olaparib
Description
Olaparib is a potent PARP2 and PARP1 inhibitor (IC50 values are 1 and 5 nM, respectively). Olaparib abolishes PARP-1 activity at a concentration of 30-100 nM in SW620 cell lysates. BRCA1-deficient cell lines are hypersensitive to PARP inhibition by Olaparib in comparison with BRCA1- and BRCA2-proficient cells. Olaparib potentiates the antitumor activity of the methylating chemotherapeutic agent Temozolomide (Cat. No. 2706) in xenograft mouse model. Olaparib is orally bioavailable in rats.
Specifications
Specifications
| Chemical Name or Material | 4-[[3-[[4-(Cyclopropylcarbonyl)-1-piperazinyl]carbonyl]-4-fluorophenyl]methyl]-1(2 H)-phthalazinone |
| CAS | 763113-22-0 |
| Quantity | 50 mg |
| Target | Poly(ADP-ribose) Polymerase Inhibitors |
| Molecular Formula | C24H23FN4O3 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?