missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Olaparib

Product Code. 17281212 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17281212 250 mg 250mg
17271212 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17281212 Supplier Thermo Scientific Chemicals Supplier No. 466292500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 763113-22-0 | C24H23FN4O3, C24H23FN4O3 | 434.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 763113-22-0
Molecular Formula C24H23FN4O3

Specifications

Quantity 250 mg
Melting Point 198.0°C
Formula Weight 434.47
InChI Key FDLYAMZZIXQODN-UHFFFAOYSA-N
SMILES FC1=C(C=C(CC2=NNC(=O)C3=CC=CC=C23)C=C1)C(=O)N1CCN(CC1)C(=O)C1CC1
IUPAC Name 4-{[3-(4-cyclopropanecarbonylpiperazine-1-carbonyl)-4-fluorophenyl]methyl}-1,2-dihydrophthalazin-1-one
Molecular Weight (g/mol) 434.47
Chemical Name or Material Olaparib
compliance-icons compliance-icons
Product Identifier
  • Olaparib
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H360D-May damage the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Olaparib

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.