missing translation for 'onlineSavingsMsg'
Learn More

Olaquindox, TRC

Product Code. 30415293
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415293 500 mg 500mg
30428007 100 mg -
30420065 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30415293 Supplier Toronto Research Chemicals Supplier No. TRCO515000500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23696-28-8
Molecular Formula C12 H13 N3 O4
Molecular Weight (g/mol) 263.25
Synonym 2-[N-(2-Hydroxyethyl)carboxamide]-3-methylquinoxaline 1,4-dioxide, Bayernox, BayoNox, Bisergon, NSC 634933, Olaquindox
IUPAC Name N-(2-hydroxyethyl)-3-methyl-1,4-dioxidoquinoxaline-1,4-diium-2-carboxamide
SMILES Cc1c(C(=O)NCCO)[n+]([O-])c2ccccc2[n+]1[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H13N3O4/c1-8-11(12(17)13-6-7-16)15(19)10-5-3-2-4-9(10)14(8)18/h2-5,16H,6-7H2,1H3,(H,13,17)
Formula Weight 263.0906
Chemical Name or Material Olaquindox
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.