missing translation for 'onlineSavingsMsg'
Learn More

Oleandomycin, TRC

Product Code. 17998394
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17998394 10 mg 10mg
17987814 1 mg 1mg
17988964 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17998394 Supplier Toronto Research Chemicals Supplier No. TRCO51750010MG
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3922-90-5
Molecular Formula C35 H61 N O12
Molecular Weight (g/mol) 687.86
Synonym Oleandomycin, Nonanoic acid, 8-(3,5-dihydroxy-2,4-dimethylhexanoyl)-5-(4-dimethylaminotetrahydro-3-hydroxy-6-methylpyran-2-yloxy)-8,9-epoxy-2,4,6-trimethyl-3-(tetrahydro-5-hydroxy-4-methoxy-6-methylpyran-2-yloxy)-, μ-lactone (6CI), 1,9-Dioxaspiro[2.13]hexadecane, oleandomycin deriv., 1,9-Dioxaspiro[2.13]hexadecane-4,10-dione, 12-[(2,6-dideoxy-3-O-methyl-α-L-arabino-hexopyranosyl)oxy]-6-hydroxy-5,7,8,11,13,15-hexamethyl-14-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-, [3R-(3R*,5R*,6S*,7R*,8R*,11R*,12S*,13R*,14S*,15S*)]-, Amimycin, Antibiotic PA 105, Landomycin, Oleandomycin A, PA 105, PA 775, Romicil, [3R-(3R*,5R*,6S*,7R*,8R*,11R*,12S*,13R*,14S*,15S*)]-12-[(2,6-Dideoxy-3-O-methyl-α-L-arabino-hexopyranosyl)oxy]-6-hydroxy-5,7,8,11,13,15-hexamethyl-14-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-1,9-dioxaspiro[2.13]hexadecane-4,10-dione
IUPAC Name (3R,5S,6S,7R,8S,9R,12R,13R,14S,15R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-hydroxy-8-[(2R,4S,5S,6S)-5-hydroxy-4-methoxy-6-methyloxan-2-yl]oxy-5,7,9,12,13,15-hexamethyl-1,11-dioxaspiro[2.13]hexadecane-10,16-dione
SMILES CO[C@H]1C[C@H](O[C@H]2[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@H](C)C[C@@]4(CO4)C(=O)[C@H](C)[C@@H](O)[C@@H](C)[C@@H](C)OC(=O)[C@@H]2C)O[C@@H](C)[C@@H]1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C35H61NO12/c1-16-14-35(15-43-35)32(40)19(4)27(37)18(3)22(7)46-33(41)21(6)31(47-26-13-25(42-11)28(38)23(8)45-26)20(5)30(16)48-34-29(39)24(36(9)10)12-17(2)44-34/h16-31,34,37-39H,12-15H2,1-11H3/t16-,17+,18-,19+,20+,21+,22+,23-,24-,25-,26-,27-,28-,29+,30-,31-,34-,35+/m0/s1
Formula Weight 687.4194
Chemical Name or Material Oleandomycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.