missing translation for 'onlineSavingsMsg'
Learn More

Oleanolic acid, 97%

Product Code. 15426918
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426918 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15426918 Supplier Thermo Scientific Alfa Aesar Supplier No. H60445.MD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 508-02-1 | C30H48O3 | 456.711 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 508-02-1
Melting Point 300°C
Molecular Formula C30H48O3
MDL Number MFCD00064914
Quantity 250 mg
Synonym oleanolic acid, oleanic acid, caryophyllin, astrantiagenin c, giganteumgenin c, virgaureagenin b, 3beta-hydroxyolean-12-en-28-oic acid, unii-6smk8r7tgj, ccris 6493, oleanolic_acid
InChI Key MIJYXULNPSFWEK-GTOFXWBISA-N
SMILES CC1(CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1)C)C(=O)O)C
IUPAC Name (4aS,6aR,6aS,6bR,8aR,10S,12aR,14bS)-10-hydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
Molecular Weight (g/mol) 456.711
PubChem CID 10494
ChEBI CHEBI:37659
Formula Weight 456.7
Percent Purity 97%
Chemical Name or Material Oleanolic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.