missing translation for 'onlineSavingsMsg'
Learn More

Oleanolic Acid, TRC

Product Code. 30011450
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011450 100 mg 100mg
30008571 500 mg 500mg
30007312 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011450 Supplier Toronto Research Chemicals Supplier No. TRCO521000100MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 508-02-1
Molecular Formula C30 H48 O3
Molecular Weight (g/mol) 456.7
Synonym Olean-12-en-28-oic acid, 3-hydroxy-, (3β)-, Olean-12-en-28-oic acid, 3β-hydroxy- (8CI), (3β)-3-Hydroxyolean-12-en-28-oic acid, (+)-Oleanolic acid, 3β-Hydroxyolean-12-en-28-oic acid, Astrantiagenin C, Caryophyllin, Giganteumgenin C, Gledigenin 1, NSC 114945, Oleanic acid, Oleanolic acid, Oleonolic acid, Virgaureagenin B
IUPAC Name (4aS,6aR,6aS,6bR,8aR,10S,12aR,14bS)-10-hydroxy-2,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
SMILES CC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2C1)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C30H48O3/c1-25(2)14-16-30(24(32)33)17-15-28(6)19(20(30)18-25)8-9-22-27(5)12-11-23(31)26(3,4)21(27)10-13-29(22,28)7/h8,20-23,31H,9-18H2,1-7H3,(H,32,33)/t20-,21-,22+,23-,27-,28+,29+,30-/m0/s1
Formula Weight 456.3603
Percent Purity >95%
Chemical Name or Material Oleanolic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.