missing translation for 'onlineSavingsMsg'
Learn More

Oleic acid, 97%

Product Code. 10455375 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10455375 25 g Glass bottle 25g
10712101 5 g Glass bottle 5g
11944961 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10455375 Supplier Thermo Scientific Acros Supplier No. 270290250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112-80-1 | C18H34O2 | 282.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
PubChem CID 445639
ChEBI CHEBI:16196
IUPAC Name (Z)-octadec-9-enoic acid
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O

Specifications

Melting Point 13.0°C
Color Colorless
Density 0.8900g/mL
Boiling Point 360.0°C
Flash Point 189°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7CH=CH(CH2)7COOH
Refractive Index 1.4585 to 1.4605
Quantity 25 g
Beilstein 02,463
Fieser 01,759
Merck Index 15,6921
Specific Gravity 0.89
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, benzene, chloroform, ether,, fixed and volatile oils
Viscosity 39.1 mPa.s (20°C)
Formula Weight 282.46
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Oleic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.