missing translation for 'onlineSavingsMsg'
Learn More

Oleic Acid, Extra Pure, SLR, Fisher Chemical™

Product Code. 10588810 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10588810 1 L 1 L
10000270 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10588810 Supplier Fisher Chemical Supplier No. O/0200/15
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

C18H34O2, CAS Number-112-80-1, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, glycon ro, glycon wo, oleate, oleic acid, pamolyn 100, wecoline oo, z-octadec-9-enoic acid, 2.5L, 360 deg.C, CHEBI:16196, Colorless, 282.46g/mol, ZQPPMHVWECSIRJ-KTKRTIGZSA-N, (Z)-octadec-9-enoic acid, 13 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
PubChem CID 445639
ChEBI CHEBI:16196
IUPAC Name (Z)-octadec-9-enoic acid
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O

Specifications

Melting Point 13°C
Color Colorless
Boiling Point 360°C
Packaging Amber glass bottle
Quantity 1 L
Vapor Density 9.7
Viscosity 39.1 mPaS at 20°C
Formula Weight 282.46g/mol
Vapor Pressure 1mmHg 176
Physical Form Liquid
compliance-icons
Product Identifier
  • Oleic acid
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.