missing translation for 'onlineSavingsMsg'
Learn More

Oleic acid, MedChemExpress

Product Code. 16472569
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472569 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16472569 Supplier Medchem Express Supplier No. HYN1446
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oleic acid (9-cis-Octadecenoic acid) is an abundant monounsaturated fatty acid. Oleic acid is a Na+/K+ ATPase activator.

Oleic acid (9-cis-Octadecenoic acid) is an abundant monounsaturated fatty acid. Oleic acid is a Na+/K+ ATPase activator.

Specifications

Chemical Name or Material Oleic acid
CAS 112-80-1
Color Colorless
Purity Grade Notes Research
Molecular Formula C18H34O2
Quantity 1 mg
Synonym 9-cis-Octadecenoic acid 9Z-Octadecenoic acid
Solubility Information Ethanol : 100 mg/mL (354.03 mM; Need ultrasonic) ∣DMSO : ≥ 62.5 mg/mL (221.27 mM)
SMILES CCCCCCCC/C=C\CCCCCCCC(O)=O
Molecular Weight (g/mol) 282.46
Formula Weight 282.46
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.