missing translation for 'onlineSavingsMsg'
Learn More

Oleic Acid, TRC

Product Code. 30411928
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411928 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411928 Supplier Toronto Research Chemicals Supplier No. TRCO524201500MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oleic Acid
CAS 112-80-1
Purity Grade Notes GC
Molecular Formula C18 H34 O2
Quantity 500 mg
Synonym Oleic acid, (Z)-Octadec-9-enoic acid, 9-Octadecenoic acid (9Z)-, 9-Octadecenoic acid (Z)-, Oleic acid (8CI), (9Z)-9-Octadecenoic acid, (18,9-Olefin) oleic acid, 9-Octadecenoic acid','(Z)-, 9-cis-Octadecenoic acid, 9Z-Octadecenoic acid, B 115, C18:1, C18:1n9c, Clear FRAC EF, Crodacid O-P, Crossential O 94, D 100, D 100 (fatty acid), Edenor ATiO5, Edenor C 18:1-98-100, Edenor FTiO5, Emersol 205, Emersol 211, Emersol 213NF, Emersol 214NF, Emersol 233, Emersol 622, Emersol 6313NF, Extra Oleic 80R, Extra Oleic 90, Extra Oleic 99, Extra Olein 80, Extra Olein 80R, Extra Olein 90, Extra Olein 90R, Extra Olein 99, Extra Olein A 1981, Impex, Industrene 105, Lubolic acid, Lunac LO-V, Lunac O-CA, Lunac O-LL, Lunac O-LL-V, Lunac O-P, Lunac O-V, Lunac OA, NAA 200, NAA 35, NAA 36, NAA 38, Neo-Fat 92-04, Oleine 7503, PM 500, PM 810B, POFAC 930, Palmera A 1813, Pamolyn 100, Priolene 6204, Priolene 6906, Priolene 6907, Priolene 6928, Priolene 6930, Priolene 6933, Priolene 6936, Radiacid 0137, Radiacid 0212, Vopcolene 27, Wecoline OO, Z-9-Octadecenoic acid, cis-9-Octadecenoic acid, cis-Oleic acid, cis-Δ9-Octadecenoic acid, Δ9-cis-Octadecenoic acid, Δ9-cis-Oleic acid
InChI Formula InChI=1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9-
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)O
IUPAC Name (Z)-octadec-9-enoic acid
Molecular Weight (g/mol) 282.46
Formula Weight 282.2559
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.